Learn More
Ohaus™ Pioneer™ PX 124M Analytical Balance
Product Code. 15805347 Shop All Ohaus Products
Click to view available options
:
Capacity: 3200 g
Product Code. 15805347
Brand: Ohaus™ 30430010
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings.
  • Competitive Performance at an Economical Price - The PX offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and educational settings at an economical pricepoint.
  • Durably Constructed for Long-Term Use - Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX is durably constructed for versatile, long-term use.
  • Designed for Simple, Intelligent Operation - The PX features a second line display for additional information or guidance, a static removal bar for convenient grounding, and USB and RS232 connectivity for easy communication.
TRUSTED_SUSTAINABILITY
Specifications
Calibration InCal™ - Semi Automatic
Certifications/Compliance CE
Connection Type RS232 & USB
Description Electronic Balance, PX124M
Capacity 120 g
Display Type Backlit LCD
Readability 0.000100 g
Languages English (US), French, German, Italian, Spanish (MX)
Linearity 0.000200 g
Material ABS Plastic, Cast Metal, Stainless Steel, Glass
Model PX124M Approved
Repeatability 0.000100 g
Stabilization Time 3.000 sec.
Weighing Modes Basic Weighing, Parts Counting, Percent Weighing, Animal Weighing, Density Determination
Dimensions (D x W x H) 321 x 209 x 309 mm
Dimensions (L x W x H) 321.00 x 209.00 x 309.000 mm
Length (Metric) 321.00 mm
Width (Metric) 209.00 mm
Height (Metric) 309.000 mm
Depth (Metric) 321.00 mm
Diameter (Metric) Outer Pan 90 mm
Weight (Metric) 4.5 kg
Weight (Metric) Shipping 7 kg
Electrical Requirements 100/240 V ∼200 mA 50/60 Hz 12/18 VA
Show More Show Less
Product Content Correction
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.