Learn More
Pindolol, MedChemExpress
Product Code. 16438546
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16438546
Brand: Medchem Express HYB0982
MedChemExpress Pindolol (LB-46) is a nonselective β-blocker with partial beta-adrenergic receptor agonist activity, also functions as a 5-HT1A receptor weak partial antagonist (Ki=33nM).
Pindolol (LB-46) is a nonselective β-blocker with partial beta-adrenergic receptor agonist activity, also functions as a 5-HT1A receptor weak partial antagonist (Ki=33nM).
Specifications
Chemical Name or Material Pindolol
CAS 13523-86-9
Color White
Purity Grade Notes Research
Molecular Formula C14H20N2O2
Quantity 100 mg
Synonym LB-46
Solubility Information DMSO : 50 mg/mL (201.35 mM; Need ultrasonic)
SMILES OC(CNC(C)C)COC1=CC=CC2=C1C=CN2
Molecular Weight (g/mol) 248.32
Formula Weight 248.32
Percent Purity 98.1%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.