Learn More
Pinaverium Bromide, TRC
Product Code. 30011038
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011038 500 mg 500mg
30009941 100 mg 100mg
30010864 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011038 Supplier Toronto Research Chemicals Supplier No. TRCP465000500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53251-94-8
Molecular Formula C26 H41 Br N O4 . Br
Molecular Weight (g/mol) 591.42
Synonym Morpholinium, 4-[(2-bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-yl)ethoxy]ethyl]-, bromide (1:1), Morpholinium, 4-[(2-bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-yl)ethoxy]ethyl]-, bromide (9CI), Dicetel, Eldicet, L-Pinaverium bromide, LAT 1717, Pinaverium bromide
IUPAC Name 4-[(2-bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethyl-4-bicyclo[3.1.1]heptanyl)ethoxy]ethyl]morpholin-4-ium;bromide
SMILES [Br-].COc1cc(Br)c(C[N+]2(CCOCCC3CCC4CC3C4(C)C)CCOCC2)cc1OC
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C26H41BrNO4.BrH/c1-26(2)21-6-5-19(22(26)16-21)7-11-31-12-8-28(9-13-32-14-10-28)18-20-15-24(29-3)25(30-4)17-23(20)27;/h15,17,19,21-22H,5-14,16,18H2,1-4H3;1H/q+1;/p-1
Formula Weight 589.1402
Percent Purity >95%
Chemical Name or Material Pinaverium Bromide
Grade HPLC