missing translation for 'onlineSavingsMsg'
Learn More

Pimobendan, Thermo Scientific™

Product Code. p-7232418 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 17382481

Brand: Thermo Scientific Acros 468572500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 74150-27-9
Molecular Formula C19H18N4O2
Molecular Weight (g/mol) 334.38
MDL Number 00761648
InChI Key GLBJJMFZWDBELO-UHFFFAOYNA-N
IUPAC Name 6-[2-(4-methoxyphenyl)-1H-1,3-benzodiazol-6-yl]-5-methyl-2,3,4,5-tetrahydropyridazin-3-one
SMILES COC1=CC=C(C=C1)C1=NC2=CC=C(C=C2N1)C1=NNC(=O)CC1C

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Concentration or Composition Notes Titration with HClO4
Physical Form Powder
Concentration 97.5% to 102.5%
UN Number 2811
Color White to Light Yellow
Merck Index 7516
Quantity 250 mg
Formula Weight 334.37
Chemical Name or Material Pimobendan
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.