Learn More
Pilocarpine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Pilocarpine Hydrochloride |
| CAS | 54-71-7 |
| Molecular Formula | C11 H16 N2 O2 . Cl H |
| Quantity | 1 g |
| Synonym | Pilocarpine Hydrochloride, (3S,4R)-3-Ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-2(3H)-furanone monohydrochloride, (3S-cis)-3-Ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-2(3H)-furanone monohydrochloride, Pilocarpine monohydrochloride, (+)-Pilocarpine hydrochloride, Adsorbocarpine, Almocarpine, Isopto Carpine, NSC 5746, Pilocar SMP, Pilocarpal, Pilocarpine chloride, Pilocarpine hydrochloride, Pilocarpine muriate, Pilocarpinium chloride, Pilocel, Pilovisc, Salagen, Vistacarpin N |
| InChI Formula | InChI=1S/C11H16N2O2.ClH/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;/h5,7-8,10H,3-4,6H2,1-2H3;1H/t8-,10-;/m0./s1 |
| SMILES | Cl.CC[C@H]1[C@@H](Cc2cncn2C)COC1=O |
| IUPAC Name | (3S,4R)-3-ethyl-4-[(3-methylimidazol-4-yl)methyl]oxolan-2-one;hydrochloride |
| Molecular Weight (g/mol) | 244.72 |
| Formula Weight | 244.0979 |
Product Title
Haben Sie Verbesserungsvorschläge?