missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3IP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113261
This item is not returnable.
View return policy
Description
PIK3IP1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
The exact function of PIK3IP1 remains unknown.
Specifications
| PIK3IP1 | |
| Polyclonal | |
| Unconjugated | |
| Pik3ip1 | |
| 1500004A08Rik; 5830455E04Rik; calcineurin-regulated kringle domain protein; calcineurin-regulated kringle domain-containing protein; Crkd; HGFL; HGFL protein; hHGFL(S); Kringle domain-containing protein HGFL; phosphoinositide-3-kinase interacting protein 1; phosphoinositide-3-kinase-interacting protein 1; Pik3ip1; RGD1311203; zgc:66482 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 113791, 216505 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.48 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q7TMJ8, Q96FE7 | |
| Pik3ip1 | |
| Recombinant Human Phosphoinositide-3-kinase-interacting protein 1 protein (22-168AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction