missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19006-1-AP-150UL
This item is not returnable.
View return policy
Description
PIF1 is a single-stranded (ss) DNA-dependent ATPase as well as a DNA helicase that unwinds DNA in a 5′ to 3′ direction with respect to the DNA strand on which it binds first. This protein is critical to DNA replication and telomere length maintece in Saccharomyces cerevisiae. The PIF1 gene is highly conserved from yeast to humans. Mutations in the gene that encodes for PIF1 cause all telomeres to lengthen, suggesting that the PIF1 functions as a catalytic inhibitor of both new telomere formation and telomere elongation. Human PIF1 preferentially binds telomeric DNA where it inhibits telomerase activity. PIF1 specifically counteracts the RecQ homolog Sgs1 helicase activity, and degradation of PIF1 is mediated by the ubiquitin-26S proteasome pathway.Specifications
| PIF1 | |
| Polyclonal | |
| Unconjugated | |
| PIF1 | |
| C15orf20, PIF, PIF1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 208084, 367645, 80119 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q1HG60, Q80SX8, Q9H611 | |
| Pif1 | |
| PIF1 Fusion Protein Ag13522 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction