Learn More
Thermo Scientific™ Tris/EDTA
Product Code. 10300924 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10300924 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10300924 Supplier Thermo Scientific™ Supplier No. 17890
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Consitently purify and store DNA without any worries using this highly purified, sterile, DNAse-free TE buffer (pH 8.0).
Thermo Scientific™ Pierce Tris-EDTA (TE) Solution is a sterile reagent for DNA purification and storage.
Additional Information and Online Ordering
Thermo Scientific Pierce Tris-EDTA (TE) Solution is a sterile buffer (pH 8.0) prepared from highly purified reagents and free of DNase activity for DNA purification and storage.
Highlights:
  • Prepared with the finest grade chemicals in specially purified water treated to remove particulates, salts, dissolved trace metals, organic compounds and microorganisms
  • Autoclave-sterilized and free of DNase contamination
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
pH 8 pH
Product Type Tris/EDTA
Quantity 1 L
Form Liquid