Learn More
Thermo Scientific™ Pierce™ Streptavidin Coated Plates
Product Code. 10195423 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Clear
White
Format:
Standard
Strip Plate
Plate Blocking:
Blocker™ BSA Buffer
SuperBlock™ Buffer
Quantity:
5 Plates
5 x 5 Plates
Unit Size:
Pack of 25
Pack of 5
Product Code. Color Format Plate Blocking Quantity unitSize
10195423 Clear Strip Plate SuperBlock™ Buffer 5 x 5 Plates Pack of 25
10485775 Black Standard Blocker™ BSA Buffer 5 Plates Pack of 5
10278243 Black Standard SuperBlock™ Buffer 5 Plates Pack of 5
10412905 Black Standard SuperBlock™ Buffer 5 Plates Pack of 5
10687764 Clear Strip Plate Blocker™ BSA Buffer 5 Plates Pack of 5
10185423 Clear Strip Plate SuperBlock™ Buffer 5 Plates Pack of 5
10155473 Clear Standard Blocker™ BSA Buffer 5 Plates Pack of 5
10288243 Clear Standard SuperBlock™ Buffer 5 Plates Pack of 5
10485005 Clear Standard SuperBlock™ Buffer 5 x 5 Plates Pack of 25
10678534 Clear Standard SuperBlock™ Buffer 5 Plates Pack of 5
10308783 White Standard Blocker™ BSA Buffer 5 Plates Pack of 5
10722984 White Standard SuperBlock™ Buffer 5 Plates Pack of 5
12 options
Product Code. 10195423
Brand: Thermo Scientific™ 15122
Thermo Scientific™ Pierce Streptavidin Coated Plates are pre-blocked and ready-to-use polystyrene 96-well and 384-well plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Streptavidin-coated microplates provide the biotin-binding function needed for a variety of ELISA and other assay procedures based on biotinylated capture antibodies, peptide antigens, and other analytes such as oligonucleotides. Pierce Streptavidin Coated Plates are available in clear, white, and black plates for use with colorimetric, chemiluminescent, and fluorescent detection methods, respectively.
Thermo Scientific Pierce Streptavidin Coated Plates are pre-blocked and ready-to-use polystyrene plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Features of Streptavidin Coated Plates include:
• Approximate binding capacity is ∼5 pmol biotin/well
• Easy and gentle immobilization of biotin-containing conjugates
• Low nonspecific binding
• No denaturing of the protein component of a conjugate upon binding
• Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene
• Pre-blocked with your choice of Blocker BSA or SuperBlock Blocking Buffer
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Streptavidin is the most popular and widely available biotin-binding protein for research methods. Streptavidin has an isoelectric point of 5 to 6, resulting in low nonspecific interactions.
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Standard Capacity, Affinity
Format Strip Plate
Content And Storage Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.
Description Clear, 8-well Strip (5 pack)
Detection Method Colorimetric
Product Line Pierce
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Surface Treatment Streptavidin
Plate Blocking SuperBlock™ Buffer
Quantity 5 x 5 Plates