Learn More
Thermo Scientific™ Pierce™ Glutathione Coated Plates
Product Code. 10492895 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Clear
White
Format:
Standard
Strip Plate
Detection Method:
Chemiluminescence
Colorimetric
Fluorescence
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Format Detection Method Unit Size
10492895 White Standard Chemiluminescence Pack of 5
10237673 Black Standard Fluorescence Pack of 5
10465775 Clear Strip Plate Colorimetric Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10492895 Supplier Thermo Scientific™ Supplier No. 15240
  • ONLINE EXCLUSIVE PRICE
    2425.00 NOK
    2109.75 NOK / Pack of 5
    Save 315.25 NOK
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-12-2026
Thermo Scientific™ Pierce Glutathione Coated Plates are standard 96-well microplates coated with intact glutathione (GSH) and pre-blocked for use in capture and detection assays involving glutathione-S-transferase (GST) fusion proteins.
Pierce Glutathione Coated Plates are coated with intact glutathione (GSH) and pre-blocked for use in capture and detection assays involving glutathione-S-transferase (GST) fusion proteins. These coated plates can be used in the analysis of cell lysates to determine the presence and concentration of the GST-fusion protein produced in an expression system. Plates can also be used to capture and coat fusion proteins via their GST tag for subsequent binding assays involving the recombinant protein. As the plates are pre-blocked, initial purification of the cell lysates is not necessary.
Thermo Scientific Pierce Glutathione Coated Plates are microplates coated with intact glutathione (GSH) and pre-blocked for use in capture and detection assays involving glutathione-S-transferase (GST) fusion proteins.
Features of Glutathione Coated Plates include:
• Selective—specifically binds fusion proteins containing glutathione S-transferase
• Capacity—binds approx. 10 ng GST protein per well
• Versatile—ready-to-bind plate format is useful for protein interaction studies
• Compatible—detergents used to lyse cells do not inhibit GST protein binding to pre-coated plates (by contrast with direct coating methods with polystyrene plates)
• Ready to use—Pre-blocked with SuperBlock Blocking Buffer to reduce nonspecific binding
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Standard Capacity, Affinity
Format Standard
Content And Storage Store at 4°C in unopened pouches. Once opened, store unused plates in a resealable bag with desiccant at 4°C.
Detection Method Chemiluminescence
Packaging 5 Plates
Product Line Pierce
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 96
Surface Treatment Glutathione
Plate Blocking SuperBlock™ Buffer
Quantity 5 plates
Show More Show Less
My initial primary culture is contaminated, what can I do?
Contamination of primary tissue could possibly be carried over to culture preparation. Change the media and treat with antibiotics to eliminate the contaminants. To prevent this in the future, wash tissue pieces several times in a balanced salt solution containing a higher concentration of antibiotics and antimycotics before starting culture.
What is the detection limit of proteins with Pierce Glutathione Coated Plates?
The background is extremely low when using the Pierce Glutathione Coated Plates. This results in a high signal-to-noise ratio and a low detection limit which in general means that the detection limit (OD 0.5) is 3 ng per well (100 µL).
How long are Pierce Glutathione Coated Plates stable?
The Pierce Glutathione Coated Plates are extremely stable over long periods of time, if stored at room temperature (20-25 degrees C). We guarantee 12 mths of shelf life in unopened foil package after the date of shipment.
Can I determine the concentration of an unknown GST-tagged protein?
To determine the approximate concentration of a GST-tagged protein, a standard curve should be generated using purified GST added in different concentrations to the wells of the plates. Please note that it is only the approximately concentration that can be determined since not 100% of the GST-fusion proteins will be bound due to denaturing or steric hindrance to GSH.
Can I whirl-mix solutions containing GST proteins?
No. Do not whirl-mix a solution containing GST-tagged proteins. The CV will increase due to slight degradation of the GST protein.
With Pierce Glutathione Coated Plates, why should I use Tween-20 surfactant in the washing step and in the buffers containing the antibodies?
We recommend using 0.05% Tween-20 surfactant (as the non-ionic detergent) to improve the signal-to-noise ratio of the assay and thus decrease the background.
Should I optimize the concentration of the purified GST or GST-tagged protein to be used with Pierce Glutathione Coated Plates?
Yes. We recommend using a solution of the purified GST or GST-tagged protein diluted in Phosphate Buffered Saline (PBS), pH 7.2 in a concentration ranging from 0.01-1 µg/mL. During the coupling, non-ionic detergents like Tween-20 surfactant should not be present, as these will suppress the coupling.
Which type of molecules can I use with Pierce Glutathione Coated Plates?
The Pierce Glutathione Coated Plates are designed for optimal detection of glutathione-S-transferase (GST)-tagged proteins or purified GST.
Can antibiotics be used with StemFlex Medium to prevent contamination during gene editing/flow sorting experiments?
Yes. We use Antibiotic-Antimycotic (Cat. No. 15240062) for additional protection from contamination.