Description
Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.
- Password protection for stored programs
- Very last liquid droplets dispensed by repeated blow-out for maximum recovery
- Features one-touch operating-button and feather-light electronic tip ejection
- Includes plate tracker for microwell plate dispensing
- Electronic piston control and brake for precise pipetting
- Safe-cone filters to prevent contamination
- Adjustment wheel for volume setting and navigation
- Range of pipetting modes for diluting and titrating to serial dispensing
- Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese
- Certificate of accredited 3-point calibration
- RFID-enabled to support asset management
Applications:
PCR and other DNA/RNA techniques, ELISA, Protein analysis and Cell culture
Certifications
CE Marked
Warranty
2 Year
Specifications
Specifications
| Calibration Points | Single |
| Certifications/Compliance | Certificate of accredited 3-point calibration |
| Compatible Tips | Universal Design, Safetyspace, Optifit |
| Autoclavable | Not Autoclavable |
| Color | Blue |
| Disposable | Reusable |
| Number of Channels | 1 |
| Product Line | Picus NxT |
| Sterility | Non-sterile |
| Type | Electronic Pipette |
| Show More |