Description
- Enhanced grip capabilities with dipped fingertips and a PVC-dotted palm
- Ideal for light and delicate applications in dry environments
- Ensures tactility and dexterity for precise and safe handling
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, REACH Compliant |
| Coating Material | PVC Dots |
| Color | Gray |
| Exterior Finish | Partially Textured |
| Hand | Specific |
| Latex Free | No |
| Material | Polyamide |
| Product Type | Mechanical Protection Gloves |
| Size | 6 |
| EN388 Standard | 2121A |
| Show More |