Learn More
Ansell Picolon Confort Gloves
Description
- Stretchable seamless liner for great fit and dexterity
- Dots on palm and fingers for enhanced grip properties
- Good resistance to abrasion and small blade cut hazards
- Lint free glove
Specifications
Specifications
| Certifications/Compliance (Europe) | CE Number, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, Category II, REACH Compliant |
| Chlorinated | No |
| Coated Area | Palm |
| Coating Material | PVC Dots |
| Color | White |
| Exterior Finish | Partially Textured |
| Hand | Specific |
| Latex Free | Yes |
| Material | PVC |
| Product Type | PVC-coated Industrial Glove |
| Show More |