missing translation for 'onlineSavingsMsg'
Learn More

Piclidenoson, MedChemExpress

Product Code. p-200101432
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16455768 10 mg 10mg
16438447 5 mg 5mg
16433376 25 mg 25mg
16401927 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16455768

Brand: Medchem Express HY13591

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Piclidenoson (IB-MECA) is a first-in-class, orally active and selective A3 adenosine receptor (A3AR) agonist. Piclidenoson exhibits antiproliferative effect and induces apoptosis in different cancer cell types like melanoma, leukemia. Piclidenoson can be used for the research of autoimmune inflammatory diseases and COVID-19.

Piclidenoson (IB-MECA) is a first-in-class, orally active and selective A3 adenosine receptor (A3AR) agonist. Piclidenoson exhibits antiproliferative effect and induces apoptosis in different cancer cell types like melanoma, leukemia. Piclidenoson can be used for the research of autoimmune inflammatory diseases and COVID-19.

Specifications

Chemical Name or Material Piclidenoson
CAS 152918-18-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H19IN6O4
Quantity 10 mg
Synonym IB-MECA CF-101
Solubility Information DMSO : ≥ 45 mg/mL (88.19 mM)
SMILES O[C@H]1[C@H](N2C=NC3=C(NCC4=CC=CC(I)=C4)N=CN=C23)O[C@H](C(NC)=O)[C@H]1O
Molecular Weight (g/mol) 510.29
Formula Weight 510.29
Percent Purity 99.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.