Learn More
Gilac™ Picking Bin
Description
- Open fronted picking bins.
- Contents easily accessible.
- Stackable up to 2 metres.
- Stability reinforced through bin feet interlocking.
- Available in 6 colors, making it easier to differentiate the contents.
Specifications
Specifications
| Type | Storage Bin |
| Material | HDPE |
| Color | White |
| Shape | Rectangular |