Learn More
Gilac™ Picking Bin
Product Code. 17948951
Change view
Click to view available options
Color:
Blue
Gray
Green
Red
White
Yellow
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17948951 White Each
17968691 Yellow Each
17948601 Blue Each
17948691 Red Each
17988991 Green Each
17928441 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17948951 Supplier Gilac™ Supplier No. G152821
  • ONLINE EXCLUSIVE PRICE
    519.75 NOK
    375.88 NOK / Pack of 1
    Save 143.87 NOK
    27% Off
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Picking Bin is open fronted for easily accessible contents. 6 available colors.
  • Open fronted picking bins.
  • Contents easily accessible.
  • Stackable up to 2 metres.
  • Stability reinforced through bin feet interlocking.
  • Available in 6 colors, making it easier to differentiate the contents.
Specifications
Type Storage Bin
Material HDPE
Color White
Shape Rectangular