Learn More
PICK1 Mouse anti-Human, Unconjugated, Clone: 2E11A4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67099-1-IG-20UL
 View more versions of this product
Product Code. 16809535
  • 1710.33 NOK / 20µL
Description
The PICK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PICK1. This antibody recognizes human, mouse, rat, pig antigen. The PICK1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PICK1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, ELISA
2E11A4
Unconjugated
20 μL
9463
IgG2a