Learn More
Hanna Instruments™ PICCOLO™ plus pH/Temperature Tester with 6.3 in. Probe
Delivers a strong signal to the meter. Hanna Instruments™ PICCOLO™ plus pH/Temperature Tester with 6.3 in. Probe features a 4-in-1 interchangeable electrode that is inexpensive, rugged and houses the pH/temperature sensors, reference system and amplifier module.
Brand:  Hanna Instruments™ HI98113
Additional Details : Weight : 1.00000kg
Product Code. 10304631
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
The PICCOLO™ plus is a lightweight digital tester that is easy to use with a replaceable probe. Conventional pH meters are susceptible to the weak, high impedance signal which makes the electrode, connector, cable and meter vulnerable to noise, humidity and dirty environments. PICCOLO™ Plus has overcome these problems with a revolutionary interchangeable amplified electrode.
  • Replaceable 6.3 in. Pre-amplified pH Electrode
  • ±0.01 pH Accuracy
  • Temperature readout
  • Automatic Temperature Compensation
  • Easy two-point manual calibration
  • 100 hour battery life
  • Simple to use
  • Economical
Specifications
Specifications
±0.01
HI1295 electrode, pH 4.01 and pH 7.01 buffer solution sachets, calibration screwdriver, batteries, rugged carrying case and instructions
1 to 13
2
32°F to 158°F
265 x 29 x 15 mm (10.4 x 1.1 x 0.6 in.)
2.5 oz.
HI1295 amplified electrode with temperature sensor
Automatic from 0 to 70°C (32 to 158°F)
0°C to 50°C
pH Temperature Stick
6.3 in. Probe
±1°C
Manual
0.01
0°C to 70°C
0.1°C
70 g
1 Year (6 Months Probe)
95% max
32°F to 122°F
3 x 1.5V
100 hr.
160 mm Probe