missing translation for 'onlineSavingsMsg'
Learn More

PIBF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14413-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16893034

  • 1611.84 NOK / 20µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein that is induced by the steroid hormone progesterone and plays a role in the maintece of pregcy. The encoded protein regulates multiple facets of the immune system to promote normal pregcy including cytokine synthesis, natural killer (NK) cell activity, and arachidonic acid metabolism. Low serum levels of this protein have been associated with spontaneous pre-term labor in humans. This protein may promote the proliferation, migration and invasion of glioma.
Specifications

Specifications

PIBF1
Polyclonal
Unconjugated
PIBF1
C13orf24, CEP90, KIAA1008, PIBF, PIBF1, RP11 505F3.1
Rabbit
Antigen Affinity Chromatography
RUO
10464, 306104, 52023
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WXW3
Pibf1
PIBF1 Fusion Protein Ag5755
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers