Learn More
PIANO Naphthalene Standard
Specifications
Specifications
| Chemical Name or Material | (1 alpha,2 alpha,3 alpha)-1,2,3-trimethylcyclopentane∣(1 alpha,2 alpha,4 alpha)-1,2,4-trimethylcyclopentane∣(1 alpha,2 alpha,4 beta)-1,2,4-trimethylcyclopentane∣(1 alpha,2 beta,3 alpha)-1,2,3-trimethylcyclopentane∣(1 alpha,2 beta,4 alpha)-1,2,4-Trimethylcyclohexane∣(1 alpha,2 beta,4 alpha)-1,2,4-trimethylcyclopentane∣(1 alpha,2 beta,4 beta)-1,2,4-Trimethylcyclohexane∣1,1,2-Trimethylcyclohexane∣1,1,4-Trimethylcyclohexane∣1,1-Dimethylcyclopentane∣1-Ethyl-1-methylcyclopentane∣cis-1,2-Dimethylcyclohexane∣cis-1,3,5-Trimethylcyclohexane∣cis-1,3-Dimethylcyclopentane∣Cyclohexane∣Cyclopentane∣Ethylcyclopentane∣Isobutylcyclohexane∣Isobutylcyclopentane∣Isopropylcyclohexane∣Isopropylcyclopentane∣Methylcyclohexane∣Methylcyclopentane∣n-Butylcyclopentane∣n-Propylcyclopentane∣trans-1,2-Dimethylcyclohexane∣trans-1,2-Dimethylcyclopentane∣trans-1,3-Dimethylcyclopentane∣trans-1,4-Dimethylcyclohexane∣trans-1-Methyl-2-(4-methylpentyl)cyclopentane∣trans-1-Methyl-2-propylcyclohexane |
| CAS | 2613-69-6, 2613-72-1, 4850-28-6, 19374-46-0, 7667-59-6, 16883-48-0 |
| Analyte or Component Names | (1 alpha,2 alpha,3 alpha)-1,2,3-trimethylcyclopentane∣(1 alpha,2 alpha,4 alpha)-1,2,4-trimethylcyclopentane∣(1 alpha,2 alpha,4 beta)-1,2,4-trimethylcyclopentane∣(1 alpha,2 beta,3 alpha)-1,2,3-trimethylcyclopentane∣(1 alpha,2 beta,4 alpha)-1,2,4-Trimethylcyclohexane∣(1 alpha,2 beta,4 alpha)-1,2,4-trimethylcyclopentane∣(1 alpha,2 beta,4 beta)-1,2,4-Trimethylcyclohexane∣1,1,2-Trimethylcyclohexane∣1,1,4-Trimethylcyclohexane∣1,1-Dimethylcyclopentane∣1-Ethyl-1-methylcyclopentane∣cis-1,2-Dimethylcyclohexane∣cis-1,3,5-Trimethylcyclohexane∣cis-1,3-Dimethylcyclopentane∣Cyclohexane∣Cyclopentane∣Ethylcyclopentane∣Isobutylcyclohexane∣Isobutylcyclopentane∣Isopropylcyclohexane∣Isopropylcyclopentane∣Methylcyclohexane∣Methylcyclopentane∣n-Butylcyclopentane∣n-Propylcyclopentane∣trans-1,2-Dimethylcyclohexane∣trans-1,2-Dimethylcyclopentane∣trans-1,3-Dimethylcyclopentane∣trans-1,4-Dimethylcyclohexane∣trans-1-Methyl-2-(4-methylpentyl)cyclopentane∣trans-1-Methyl-2-propylcyclohexane |
| Shipping Condition | Room Temperature |
| Solution Type | Mixture |
| Solvent or Matrix | Hydrocarbons |
| Industry Type | Petrochemical Testing |
| Quantity | 0.1 mL |
| Concentration or Composition (by Analyte or Components) | cis-1,3-Dimethylcyclopentane 0.5899 wt%∣(1 alpha,2 alpha,3 alpha)-1,2,3-trimethylcyclopentane 0.7934 wt%∣1-Ethyl-1-methylcyclopentane 1.0432 wt%∣trans-1,2-Dimethylcyclopentane 1.4533 wt%∣(1 alpha,2 beta,3 alpha)-1,2,3-trimethylcyclopentane 1.5655 wt%∣(1 alpha,2 beta,4 alpha)-1,2,4-trimethylcyclopentane 1.6244 wt%∣trans-1,2-Dimethylcyclohexane 1.6319 wt%∣trans-1,3-Dimethylcyclopentane 2.7176 wt%∣Methylcyclopentane 3.2404 wt%∣1,1,2-Trimethylcyclohexane 3.2927 wt%∣1,1-Dimethylcyclopentane 3.4162 wt%∣Isopropylcyclopentane 3.4589 wt%∣(1 alpha,2 beta,4 alpha)-1,2,4-Trimethylcyclohexane 3.4626 wt%∣cis-1,3,5-Trimethylcyclohexane 3.4813 wt%∣Ethylcyclopentane 3.5083 wt%∣(1 alpha,2 beta,4 beta)-1,2,4-Trimethylcyclohexane 3.5637 wt%∣n-Propylcyclopentane 3.5954 wt%∣n-Butylcyclopentane 3.6004 wt%∣trans-1,4-Dimethylcyclohexane 3.6018 wt%∣1,1,4-Trimethylcyclohexane 3.631 wt%∣Isobutylcyclopentane 3.6649 wt%∣cis-1,2-Dimethylcyclohexane 3.6679 wt%∣(1 alpha,2 alpha,4 beta)-1,2,4-trimethylcyclopentane 3.6734 wt%∣(1 alpha,2 alpha,4 alpha)-1,2,4-trimethylcyclopentane 3.6982 wt%∣trans-1-Methyl-2-propylcyclohexane 3.7034 wt%∣trans-1-Methyl-2-(4-methylpentyl)cyclopentane 3.7326 wt%∣Cyclopentane 4.8512 wt%∣Cyclohexane 5.2581 wt%∣Methylcyclohexane 5.5375 wt%∣Isopropylcyclohexane 5.6489 wt%∣Isobutylcyclohexane 5.7953 wt% |