Learn More
PI3 Kinase p110 Beta Mouse anti-Human, Unconjugated, Clone: 1B2A1, Proteintech
Product Code. 16899565 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16899565 150 μL 150µL
16809575 20 μL 20µL
2 options
Product Code. 16899565
Brand: Proteintech 671211IG150UL
Mouse Monoclonal Antibody
The PI3 Kinase p110 Beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PI3 Kinase p110 Beta. This antibody recognizes human, rat antigen. The PI3 Kinase p110 Beta antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Antigen PI3 Kinase p110 Beta
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 1B2A1
Concentration 860 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 5291
Target Species Human, Rat
Isotype IgG1