missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI 3 Kinase p55 gamma Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27035-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
PIK3R3 binds to activated (phosphorylated) protein-tyrosine kinases through its SH2 domain and regulates their kinase activity. During insulin stimulation, it also binds to IRS-1. It is a component of a heterodimer of p110 (catalytic) and p55 (regulatory) subunits. The protein is expressed at highest levels in brain and testis. Lower levels are detected in adipose tissue, kidney, heart, lung and skeletal muscle. The protein contains 2 SH2 domains.Specifications
PI 3 Kinase p55 gamma | |
Polyclonal | |
Unconjugated | |
PIK3R3 | |
p55, p55 GAMMA, p55PIK, PI3 kinase subunit p55 gamma, PI3K regulatory subunit gamma, PIK3R3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8503 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q92569 | |
PIK3R3 | |
PI 3 Kinase p55 gamma Fusion Protein Ag25884 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |