Learn More
Phytolaccagenin, TRC
Product Code. 30012896
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012896 2 mg 2mg
30005656 1 mg 1mg
30012277 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012896 Supplier Toronto Research Chemicals Supplier No. TRCP3994782MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1802-12-6
Molecular Formula C31H48O7
Molecular Weight (g/mol) 532.71
Synonym Olean-12-ene-28,30-dioic acid, 2beta,3beta,23-trihydroxy-, 30-methyl ester (8CI), Phytolaccagenin (7CI), Jaligonic acid 30-methyl ester, NSC 116453, Olean-12-ene-28,29-dioic acid, 2,3,23-trihydroxy-, 29-methyl ester, (2beta,3beta,4alpha,20beta)-
IUPAC Name (2S,4aR,6aR,6aS,6bR,8aR,9R,10R,11S,12aR,14bS)-10,11-dihydroxy-9-(hydroxymethyl)-2-methoxycarbonyl-2,6a,6b,9,12a-pentamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
SMILES COC(=O)[C@@]1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@H](O)[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(=O)O
Specifications
Quantity 2 mg
InChI Formula InChI=1S/C31H48O7/c1-26(25(37)38-6)11-13-31(24(35)36)14-12-29(4)18(19(31)15-26)7-8-22-27(2)16-20(33)23(34)28(3,17-32)21(27)9-10-30(22,29)5/h7,19-23,32-34H,8-17H2,1-6H3,(H,35,36)/t19-,20-,21+,22+,23-,26-,27-,28-,29+,30+,31-/m0/s1
Formula Weight 532.34
Chemical Name or Material Phytolaccagenin