Learn More
Physcion, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Emodin 3-methyl ether |
| CAS | 521-61-9 |
| Molecular Formula | C16 H12 O5 |
| Quantity | 10 mg |
| Synonym | Anthraquinone','1,8-dihydroxy-3-methoxy-6-methyl- (8CI), Chrysophanic acid','6-methoxy- (4CI), 1,8-Dihydroxy-3-methoxy-6-methyl-9,10-anthracenedione, 1,8-Dihydroxy-3-methoxy-6-methyl-9,10-anthraquinone, 1,8-Dihydroxy-3-methoxy-6-methylanthraquinone, 1,8-Dihydroxy-3-methyl-6-methoxyanthraquinone, 1,8-Dihydroxy-6-methoxy-3-methyl anthraquinone, 6-O-Methylemodin, Emodin 3-methyl ether, NSC 251670, Parietin, Physcion, Physcione, Rheochrysidin |
| InChI Formula | InChI=1S/C16H12O5/c1-7-3-9-13(11(17)4-7)16(20)14-10(15(9)19)5-8(21-2)6-12(14)18/h3-6,17-18H,1-2H3 |
| SMILES | COc1cc(O)c2C(=O)c3c(O)cc(C)cc3C(=O)c2c1 |
| IUPAC Name | 1,8-dihydroxy-3-methoxy-6-methylanthracene-9,10-dione |
| Molecular Weight (g/mol) | 284.26 |
| Formula Weight | 284.0685 |