Description
- Xtra Grip aqua-polymer foam coating is breathable and provides a high level of mechanical abrasion resistance
- Enhanced grip capabilities in oily areas
- Tactile feel when assembling oily parts
- Ideal for precision work, maintenance, repair work, and metal processing
Specifications
Specifications
| Construction | Seamless Knit |
| Coating Material | Aqua-Polymer Foam |
| Color | Black, Gray |
| Exterior Finish | Smooth |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Spandex, Polyamide |
| Product Type | Mechanical Protection Gloves |
| Size | 6 |
| Style | Full-hand Mitt |
| Show More |