Learn More
uvex Phynomic X-Foam HV Safety Gloves
Product Code. p-7230503 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17200371 6 10 pairs
17210371 7 10 pairs
17220371 8 10 pairs
17230371 9 10 pairs
17240371 10 10 pairs
17250371 11 10 pairs
17260371 12 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17200371 Supplier uvex Supplier No. 6005406
Only null left
Add to basket
Add to basket
A seamless break section integrated into the fingers significantly reduces the risk of serious hand injuries when using manually operated screw drivers. Aqua-polymer foam coating on the palm and fingertips enhances grip capabilities.
  • Lightweight construction acts as a second-skin to maintain a high level of feeling
  • Aqua-polymer foam coating is moisture repellent and flexible
  • Breathable materials help prevent excessive perspiration
  • Seamless break section integrated into the fingers helps reduce serious hand injuries
  • Should the glove/finger become trapped in or near a piece of machinery, simply detach and tear along the perforated seam
  • Enhanced grip in dry and slightly damp applications
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Orange, Gray
Hand Ambidextrous
Material Polyamide, Elastane
Product Type Mechanical Protection Gloves
Size 6
EN388 Standard 31X1X
Gender Unisex
Glove Cuff Style Knit
Series Phynomic X-Foam HV