Learn More
uvex Phynomic FOAM Protective Gloves with Aqua-Polymer Foam Coating
Shop All Uvex Products
Click to view available options
Size:
5
8
9
10
12
Unit Size:
10 pairs
Each
Description
- High dexterity safety glove designed for precision mechanical work
- Damp-resistant, aqua-polymer foam coating provides mechanical abrasion resistance
- Highly-breathable palm coating enhances grip capabilities in dry and slightly damp areas
- Tactile feel when assembling small parts
- Ideal for precision assembly work, inspection, sorting, food processing, and packaging
Specifications
Specifications
| Certifications/Compliance | EN 388 (3 1 2 1 X) |
| Construction | Seamless Knit |
| Coating Material | Aqua-Polymer Foam |
| Color | White, Gray |
| Exterior Finish | Smooth |
| Hand | Ambidextrous |
| Material | Spandex, Polyamide |
| Product Type | Mechanical Protection Gloves |
| Size | 5 |
| Style | Full-hand Mitt |
| Show More |