missing translation for 'onlineSavingsMsg'
Learn More

PHOX2B Mouse anti-Human, Mouse, Clone: 1F5F5, Proteintech

Product Code. 16807965 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16807965 20 μL 20µL
16897955 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16807965

Brand: Proteintech 662541IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.

Specifications

Antigen PHOX2B
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1F5F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHOX2B
Gene Accession No. O35690, Q99453
Gene Alias NBPhox, Neuroblastoma Phox, paired like homeobox 2b, PHOX2B, PHOX2B homeodomain protein, PMX2B
Gene Symbols Phox2b
Host Species Mouse
Immunogen PHOX2B Fusion Protein Ag13196
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18935, 8929
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.