missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHOX2A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25804-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
PHOX2A and PHOX2B (Paired mesoderm homeobox protein) are closely related, paired-homeodomain transcription factors that function as determits of the noradrenergic phenotype during embryogenesis. PHOX2 proteins are crucial for the regulation of endogenous hydroxylases in neural crest cells and promote sympathetic neuron generation. Human PHOX2B contains one DNA binding homeobox domain and is required for the differentiation of all central and nonperipheral noradrenergic centers in the brain. In contrast, PHOX2A controls only the differentiation of the main noradrenergic center of the brain. Regulation of PHOX2 may have therapeutic utility in aging or disorders involving degeneration of noradrenergic neurons.Specifications
PHOX2A | |
Polyclonal | |
Unconjugated | |
PHOX2A | |
ARIX, ARIX1 homeodomain protein, CFEOM2, FEOM2, paired like homeobox 2a, PHOX2A, PMX2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
401 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14813 | |
PHOX2A | |
PHOX2A Fusion Protein Ag22873 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |