Learn More
Lovibond™ Photometry Test Reagent
Product Code. 11785428
Click to view available options
Quantity:
100 Pack
15 mL
Type:
Triazole Reagent, F25
Urea Reagent 2
Unit Size:
15mL
Each
Pack of 100
Product Code. 11785428
Brand: Lovibond™ 459300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Lovibond™ Photometry Test Reagent is manufactured under tightly controlled conditions to maintain the highest quality standards.
  • Highest quality production standards
  • Maximum accuracy
Specifications
Format Liquid
Quantity 15 mL
Type Urea Reagent 2