Learn More
Palintest™ Photometer Tablet Reagents
Product Code. 11993607
Change view
Click to view available options
Range:
0 to 0.5mg/L
0 to 1.5mg/L
0 to 100mg/L
0 to 2.5mg/L
0 to 20mg/L
0 to 250mg/L
0 to 500mg/L
2.5 to 2mg/L
Type:
Alkalinity Total (Alkaphot) Test Kit
Chlorine Dioxide HR Test Kit
Chlorine Dioxide LR Test Kit
Chlorine HR Test Kit
Fluoride Test Kit
Molybdate HR Test Kit
Organophosphonate (OP) Test Kit
Sulphide Test Kit
Unit Size:
50 tests
Each
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Type unitSize
11993607 0 to 250mg/L Chlorine HR Test Kit Pack of 50
12581457 0 to 2.5mg/L Chlorine Dioxide LR Test Kit Pack of 50
13108840 2.5 to 2mg/L Chlorine Dioxide HR Test Kit 50 tests
11903627 0 to 1.5mg/L Fluoride Test Kit Pack of 50
13151451 0 to 20mg/L Organophosphonate (OP) Test Kit Each
11923657 0 to 500mg/L Alkalinity Total (Alkaphot) Test Kit Pack of 50
11933627 0 to 100mg/L Molybdate HR Test Kit Pack of 50
11913657 0 to 0.5mg/L Sulphide Test Kit Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11993607 Supplier Palintest™ Supplier No. PM162
Photometer reagents in convenient tablet form. Palintest™; Photmoeter Tablet Reagents are available for a variety of water testing needs.
  • Convenient tablet form.
  • Easy to use.
  • Available for a variety of water testing needs.
Specifications
No. of Tests 50
Quantity 50/Pk.
Range 0 to 250mg/L
Sufficient For 50 Tests
Type Chlorine HR Test Kit
compliance-icons
Product Identifier
  • TEST CHLORE TOTAL 0-250MG/L
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash contaminated skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: adipic acid, citricacid, potassium iodide