Learn More
Brady™ BradyGlo™ Photoluminescent Polyester Glow-in-the-dark Warning Tape
Product Code. 11701965
Click to view available options
Width (Metric):
25.4 mm
50.8 mm
Unit Size:
1 roll
Product Code. 11701965
Brand: Brady™ 224072
BradyGlo™ B-324 Warning Tape exceeds ASTM E 2072-00 standard specifications and standards for 10+ hour glow requirements.
  • Exceeds ASTM E 2072-00 standard specifications as well as the following standards for 10+ hour glow requirements: ASTM/E162/E648/E662, SMP800C, IMO Resolution A. 752(18), ISO/CD 15370, PSPA Standard 002 Part 2 1993 Class A Rev. 2: 09/99, DIN 67 510, Parts 1-4, NFPA 101 Life Safety Code 2000 7.10.7.2, OSHA 1910.37 and Lloyd's register Cert. 01/6007
Specifications
Certifications/Compliance ASTM E–2072-00 / ASTM E–2073-00, ASTM E-162, ASTM E-648, ASTM E-662, IMO Resolution A.752(18), ISO/CD 15370, Marine Safety Committee MSC.27(61), DIN 67 510 (Parts 1 - 4), and PSPA Standard 002 Part 2
Description Glow-in-the-dark warning tape
Adhesive Type Acrylic, Pressure-sensitive, Permanent
Coating None
Color Photoluminescent
Display Blank
Finish Matte
Format Roll
Material Polyester
Printable Sides Single-side Printable
Print Technology None
Product Type Photoluminescent Tape
Sterility Non-sterile
For Use With (Application) For exit and directional signs, identification of fire alarms and extinguishers, low location egress pathway markings, and directional systems for escape or evacuations routes. Recommended Uses of Photoluminescent Safety Markings
Gloss 60° 121 Gardner Units
Min. Operating Temperature (Metric) -40°C
No. per Pack 1 Roll
Quantity 1 Ea.
Temperature Range (English) -40°F to +176°F
Temperature Range (Metric) -40°C to +80°C
Thickness (English) 0.0079 in.
Thickness (Metric) 0.201 mm
Dimensions (LxW) 9.14 m x 50.80 mm
Length (English) 10 yd.
Length (Metric) 9.14 m
Width (English) 2 in.
Width (Metric) 50.8 mm
Width (English) Printable Area 2 in.
Height (English) 1.02 in.
Height (Metric) 26 mm