Learn More
WTW™ photoLab™ S12 Filter Photometers
Ideal for comprehensive routine measuring operations in wastewater and drinking water, food industry as well as in service and teaching labs. The photoLab™ S12 filter photometer with 12 wavelengths combines lab precision with highest comfort and highest speeds for extensive water analytics
Brand:  WTW™ 250024D
Additional Details : Weight : 2.70900kg
Product Code. 11775763
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • The photoLab™ S12 filter photometer with 12 wavelengths combines lab precision with highest comfort and highest speeds for extensive water analytics
  • photoLab™ S12 with reference beam optics has 12 wavelengths for quick tests in 16 mm round and rectangular cuvettes. 
  • Barcode detection for round and rectangular cuvettes
  • AQA/IQC multi-level
  • User-defined methods, easy to operate and maintenance-free
  • Maximum comfort at minimal costs! Just plug in the cuvette, read measured value, done
  •  
Specifications
Specifications
LCD
10.62 in.
5.51 in.
50 Hz
340, 410, 445, 500, 525, 550, 565, 605, 620, 665, 690, 820nm
10.23 in.
±2nm
270 mm
230 V
140 mm
260 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products