Learn More
Thermo Scientific™ Phosphodiesterase 4 Inhibitor
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Pyrazole compound with a high-affinity active site binding inhibitor of phosphodiesterases IVB and IVD.
Specifications
Specifications
| Quantity | 5 mg |
| Description | Pale Yellow |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Pyrazole compound with a high-affinity active site binding inhibitor of phosphodiesterases IVB and IVD |
| CAS | 346440-86-6 |
| Molecular Formula | C14H15N3O4 |
| Formula Weight | 289.29g/mol |
| Synonym | Ethyl 3,5-dimethyl-1-(3-nitrophenyl)-1H-pyrazole-4-carboxylate |
| Sensitivity | Light Sensitive |
| Product Type | Signal Transduction Reagent |
| Show More |