Learn More
Phospho-PKC Alpha (Ser657) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28926-1-AP-20UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17541386
  • 1496.71 NOK / 20µL
Estimated Shipment: 02-05-2024
Description
The Phospho-PKC Alpha (Ser657) antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human Phospho-PKC Alpha (Ser657). This antibody recognizes human antigen. The Phospho-PKC Alpha (Ser657) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
Phospho-PKC Alpha (Ser657)
Polyclonal
Rabbit
Primary
Human
Western Blot, ELISA
Unconjugated
20 μL
5578