missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-P53 (Ser392) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28963-1-AP-100UL
This item is not returnable.
View return policy
Description
Phospho-P53 (Ser392) Polyclonal antibody specifically detects Phospho-P53 (Ser392) in Human samples. It is validated for WB, ELISASpecifications
| Phospho-P53 (Ser392) | |
| Polyclonal | |
| Unconjugated | |
| BC113422 | |
| Antigen affinity purification | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, ELISA | |
| 260 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Rabbit | |
| 100 μL | |
| 7157 | |
| Store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction