Learn More
Phospho-P53 (Ser15) Mouse, anti-Human, Unconjugated, Clone: 1H6G1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67826-1-IG-100UL
Description
TP53, also known as P53 and NY-CO-13, belongs to the p53 family and has 9 isoforms. In SDS-Page, the observed molecular weight is about 53 kDa. TP53 acts as a tumor suppressor in many tumor types, including growth arrest or apoptosis depending on the physiological circumstances and cell types. It is involved in cell cycle regulation as a trans-activator that acts to negatively regulate cell division by controlling a set of genes required for this process. TP53 Localizes in the nucleus in most cells but found in the cytoplasm in some cells. (PMID: 26166714; PMID: 25225161)
Specifications
| Phospho-P53 (Ser15) | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1H6G1 | |
| Unconjugated | |
| 100 μL | |
| 7157 | |
| Liquid |