missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-GSK-3 beta (S9) Rabbit anti-Human, Clone: 1253C, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8934-SP
This item is not returnable.
View return policy
Description
GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
| GSK-3 beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK3 beta, GSK-3 beta, GSK3B, GSK3beta isoform | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2932 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 1253C | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1-25 ug/mL | |
| P49840 | |
| GSK3B | |
| Phosphopeptide containing the human GSK-3 beta S9 site | |
| 25 μg | |
| Primary | |
| Detects human GSK-3 beta when phosphorylated at S9 in Western blots. | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?