missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-EphA3/EphA4/EphA5 (Tyr779, Tyr833) Polyclonal Antibody

Product Code. 15544751
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15544751 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15544751 Supplier Invitrogen™ Supplier No. PA537813

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control for IHC is human brain tissue.

These proteins belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene encodes a protein that binds ephrin-A ligands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EphA3/EphA4/EphA5 (Tyr779, Tyr833)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPHA4
Gene Accession No. P29320, P54756, P54764
Gene Alias 2900005C20Rik; AI385584; AI854630; AW125296; AW492086; Brain-specific kinase; bsk; Cek4; CEK7; CEK-7; Cek8; eck-like sequence 1; EHK1; EHK-1; EK4; EK7; EK8; Ekh1; Els1; Els1.; End3; EPH homology kinase 1; Eph receptor A3; Eph receptor A4; EPH receptor A5; EPHA3; EPHA4; Epha5; EPH-like kinase 4; EPH-like kinase 7; EPH-like kinase 8; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; ephrin type-A receptor 4; Ephrin type-A receptor 5; ETK; ETK1; H TYRO4; HEK; Hek4; Hek7; HEK8; Human embryo kinase; human embryo kinase 1; Mek4; rb; receptor protein-tyrosine kinase HEK7; receptor protein-tyrosine kinase HEK8; rEK4; Rek7; RGD1560587; Sek; Sek1; testicular tissue protein Li 64; TYRO1; TYRO1 protein tyrosine kinase; TYRO4; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor EHK-1; tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase receptor MPK-3; tyrosine-protein kinase receptor REK4; tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase receptor SEK-1; tyrosine-protein kinase TYRO1; Tyrosine-protein kinase TYRO4
Gene Symbols EPHA3, EPHA4, Epha5
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of tyrosine 779/833 (E-A-Y(p)-T-T)/(Q-D-V(p)-I-K) derived from Human EPHA3/4/5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2042, 2043, 2044
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.