missing translation for 'onlineSavingsMsg'
Learn More

Phospho-AKT (Ser473) Mouse anti-Human, Mouse, CoraLite 594, Clone: 1C10B8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL594-66444-100UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17699956

  • 3499.43 NOK / 100µL
Estimated Shipment: 02-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Phospho-AKT (Ser473) Monoclonal antibody apecifically detetects Phospho-AKT (Ser473) antigen in Human, Mouse Samples. It is validated for Flow Cytometry.
Specifications

Specifications

Phospho-AKT (Ser473)
Monoclonal
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
AKT1
Peptide
100 μL
207
Store at -20°C. Avoid exposure to light.
IgG1
Flow Cytometry
1C10B8
CoraLite 594
NM_005163
Mouse
Protein A purification
Primary
Human, Mouse
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers