missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-AKT (Ser473) Mouse anti-Human, Mouse, CoraLite 594, Clone: 1C10B8, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 5307.92 NOK
Specifications
| Antigen | Phospho-AKT (Ser473) |
|---|---|
| Clone | 1C10B8 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | CoraLite 594 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17751419
|
Proteintech
CL594-66444-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17699956
|
Proteintech
CL594-66444-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-AKT (Ser473) Monoclonal antibody apecifically detetects Phospho-AKT (Ser473) antigen in Human, Mouse Samples. It is validated for Flow Cytometry.Specifications
| Phospho-AKT (Ser473) | |
| Flow Cytometry | |
| CoraLite 594 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 207 | |
| Peptide | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 1C10B8 | |
| Monoclonal | |
| Liquid | |
| Human, Mouse | |
| NM_005163 | |
| AKT1 | |
| IgG1 | |
| Protein A purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title