Learn More
Phosphates (PO43-) Standard for Ion Chromatography, 1000 mg/L in water, Honeywell Fluka™
Product Code. 16604244 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
16604244 100 mL 100mL
1 options
Product Code. 16604244
Brand: Honeywell Fluka E3039100ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phosphate Standard for Ion Chromatography
Density 1.01 g/cm3
Quantity 100 mL
Grade Standards for Ion Chromatography