Learn More
WTW™ Phosphate Test Kits
Click to view available options
Type:
COD O2 Cell Test Kit
Phosphate (PO4) Cell Test Kit
n-Ten (nonionic) Surfactant Cell Test Kit
No. of Tests:
25
90
95
Unit Size:
2 L
Each
Pack of 25
Description
- Variety of instrumentation for measuring phosphates
- PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, Photoflex photometers
- For waste water, drinking water, surface water, fertilizers
- CombiCheck Phosphate (PO<sub>4</sub>) cell test kit
Specifications
Specifications
| Analytes | Chromosulfuric acid oxidation |
| For Use With (Application) | For waste water, drinking water, surface water, fertilizers |
| For Use With (Equipment) | For PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex, Photoflex photometers |
| Includes | Certificates |
| No. of Tests | 95 |
| Range | 0.010 to 5 mg/L PO4-P, 0.03 to 15.3 mg/L PO4 |
| Sufficient For | 25 Tests |
| Test Method | Photometric |
| Type | COD O2 Cell Test Kit |
Safety and Handling
Product Identifier
- Phosphate Test (o-phosphate)
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : sulphuric acid