Learn More
Palintest™ Phosphate LR Starter Tdisc Test
Product Code. 11934666
Change view
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
Product Code. Quantity unitSize
11934666 50 Pack Pack of 50
1 options
Product Code. 11934666
Brand: Palintest™ CM177
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with Palintest™ comparator
The Palintest™ comparator with interchangeable colour discs is simple to use. Just add the reagent tablet to the test sample, place in the comparator and match against the appropriate disc.
Specifications
Concentration 0 to 4 mg/L
Quantity 50 Pack