Learn More
Medicago Phosphate Buffered Saline Tablets with Tween™ 20, pH 7.4
Product Code. 11300439
Change view
Click to view available options
Quantity:
100 x 1000 mL Tablets
100 x 500 mL Tablets
Unit Size:
Each
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11300439 100 x 1000 mL Tablets Pack of 100
11310439 100 x 500 mL Tablets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11300439 Supplier Medicago Supplier No. 099410100
  • ONLINE EXCLUSIVE PRICE
    3650.00 NOK
    3280.79 NOK / Pack of 100
    Save 369.21 NOK
    10% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Supplied as exactly pre-weighed tablets with a final buffer volume of 500 or 1000ml. Medicago Phosphate Buffered Saline Tablets with Tween™ 20, pH 7.4 is dissolved in deionized water to yield a solution of sodium chloride, potassium chloride, Tween™ 20 and phosphate buffer.
PBS-T buffer contains the non-ionic detergent Tween™ 20 which acts through blocking. It has the ability to reduce non-specific binding and protein-protein interaction during the wash step in protein and immunoassay procedures such as ELISA and Western blotting. Decreasing the non-specific binding and staining makes ELISA results and blots easier to interpret.
  • Formulated from analytical grade reagents
  • PBS with non-ionic detergent
  • Specifically developed for immunoassays
  • Exactly pre-weighed tablets
  • Dissolve and go for greater convenience
Specifications
For Use With (Application) ELISA
pH 7.4 at 25°C
Physical Form Tablets
Quantity 100 x 1000 mL Tablets
Formulation NaCl (0.14M), KCl (0.0027M), PO4 (0.01M), Tween™ 20 (0.05%)