missing translation for 'onlineSavingsMsg'
Learn More

Melford Phosphate Buffered Saline

Product Code. 30018685
Change view
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018685 1 L 1 L
30018686 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018685 Supplier Melford Supplier No. P320601000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PBS 10X Solution, is a widely used reagent in life science research.

PBS 10X Solution, is a widely used reagent in life science research. It is a buffered saline solution that contains phosphate ions, which help maintain a stable pH level in biological and biochemical experiments.

PBS 10X Solution is commonly used in various applications such as cell culture, immunohistochemistry, immunocytochemistry, and molecular biology techniques. The primary function of PBS 10X Solution is to provide a stable environment for biological samples by maintaining a physiological pH and ionic strength.

The phosphate buffer system present in PBS helps to resist changes in pH caused by the addition of acids or bases, making it an essential tool for experimental procedures where pH control is crucial. PBS 10X Solution is prepared as a concentrated solution, typically at 10 times the final desired concentration. This concentrated form allows researchers to easily dilute it to the required working concentration for their specific experiments. It is available commercially from various suppliers and comes in different formats, such as liquid or powder form.

The versatility and reliability of PBS 10X Solution have made it a standard reagent in many laboratory protocols. Its use in cell biology and molecular biology research ensures consistent and reproducible results. PBS 10X Solution is a common choice for washing and resuspending cells, tissues, or protein samples, as it provides the necessary osmolarity and pH stability required for these procedures.

Specifications

Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.