Learn More
Thermo Scientific Chemicals Phosphate, 0.2M buffer soln., pH 7.4
Product Code. p-7076542
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
Product Code. 15483159
Brand: Thermo Scientific Chemicals J60380.AP
Request Bulk or Custom Format
Edge
Widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. | CAS: 7558-80-7119.976 g/mol
This buffer is widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. It is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7558-80-7
Molecular Weight (g/mol) 119.976
Specifications
Concentration 0.2M buffer soln.
Physical Form Liquid
Quantity 500 mL
Packaging Plastic Bottle
Solubility Information Fully miscible in water
Chemical Name or Material Phosphate