Learn More
Thermo Scientific Chemicals Phosphate, 0.2M buffer soln., pH 4.4
Catalog Number p-7076544
Click to view available options
Quantity:
500 mL
1 L
7558-80-7
7558-80-7
Widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
This buffer is widely used in molecular biology, biochemistry and chromatography. Useful in conjunction with Sodium Phosphate, dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies. It is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Concentration 0.2M buffer soln.
Molecular Weight (g/mol) 119.976
Quantity 500 mL
Packaging Plastic Bottle
Content And Storage Keep cold
Form Liquid