missing translation for 'onlineSavingsMsg'
Learn More

Phosmet, TRC

Product Code. 30136888
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136888 5 g 5g
30136031 1 g 1g
18071730 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136888 Supplier Toronto Research Chemicals Supplier No. TRCP3537005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 732-11-6
Formula Weight 316.9945
InChI Formula InChI=1S/C11H12NO4PS2/c1-15-17(18,16-2)19-7-12-10(13)8-5-3-4-6-9(8)11(12)14/h3-6H,7H2,1-2H3
IUPAC Name 2-(dimethoxyphosphinothioylsulfanylmethyl)isoindole-1,3-dione
Molecular Formula C11 H12 N O4 P S2
Molecular Weight (g/mol) 317.32
SMILES COP(=S)(OC)SCN1C(=O)c2ccccc2C1=O
Synonym Phosphorodithioic acid, O,O-dimethyl S-phthalimidomethyl ester (6CI), Phosphorodithioic acid, O,O-dimethyl ester, S-ester with N-(mercaptomethyl)phthalimide (8CI), Decemthion, Decemthion EK 20, Decemthion P-6, Decemtion, Fosmet, Ftalophos, GWN 1976, Imidan, Imidan 70WP, Imidathion, N-(Mercaptomethyl)phthalimide S-(O,O-dimethyl phosphorodithioate), O,O-Dimethyl S-(phthalimidomethyl) dithiophosphate, O,O-Dimethyl S-phthalimidomethyl phosphorodithioate, O,O-Dimethyl S-phthalimidomethyl phosphorodithioate, Orbisect, Ordatox, PMP, PMP (pesticide), Phosmet, Phthalophos, Porect, Prolate, R 1504, S-Phthalimidomethyl O,O-dimethyl phosphorodithioate, Safidon, Simidan, Starbar GX 118, Stauffer R 1504, Phosphorodithioic acid, S-[(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl] O,O-dimethyl ester
Chemical Name or Material Phosmet

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.