missing translation for 'onlineSavingsMsg'
Learn More

PHLPPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25244-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16893815

  • 3828.53 NOK / 150µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Protein phosphatase involved in regulation of Akt and PKC signaling. Mediates dephosphorylation in the C-terminal domain hydrophobic motif of members of the AGC Ser/Thr protein kinase family; specifically acts on ′Ser-473′ of AKT1, ′Ser-660′ of PRKCB isoform beta-II and ′Ser-657′ of PRKCA. Akt regulates the balance between cell survival and apoptosis through a cascade that primarily alters the function of transcription factors that regulate pro- and antiapoptotic genes. Dephosphorylation of ′Ser-473′ of Akt triggers apoptosis and decreases cell proliferation. Also controls the phosphorylation of AKT3. Dephosphorylates STK4 on ′Thr-387′ leading to STK4 activation and apoptosis. Dephosphorylates RPS6KB1 and is involved in regulation of cap-dependent translation. Inhibits cancer cell proliferation and may act as a tumor suppressor. Dephosphorylation of PRKCA and PRKCB leads to their destabilization and degradation. Dephosphorylates RAF1 inhibiting its kinase activity.
Specifications

Specifications

PHLPPL
Polyclonal
Unconjugated
PHLPP2
KIAA0931, PHLPP like, PHLPP2, PHLPPL
Rabbit
Antigen Affinity Chromatography
RUO
23035, 244650, 498949
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6ZVD8, Q8BXA7
Phlpp2
PHLPPL Fusion Protein Ag19000
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers